Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007789.3 | NP_031815.2 | 1 - 2 | 115 | 91 | |
GenBank mRNA | BC065118.1 | - | 1 - 2 | 81 | 6853253 | 91 |
CX202471.1 | - | 1 - 2 | 115 | 91 | ||
X84727.1 | - | 1 - 2 | 77 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |