Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008548.4 | NP_032574.1 | 9 - 10 | 1848 | 79 | |
GenBank mRNA | AK031038.1 | - | 9 - 10 | 1849 | 79 | |
AK042371.1 | - | 8 - 9 | 1689 | 79 | ||
AK043932.1 | - | 9 - 10 | 1789 | 79 | ||
AK154820.1 | - | 9 - 10 | 1790 | 79 | ||
AK171841.1 | - | 9 - 10 | 1791 | 79 | ||
BC015265.1 | - | 9 - 10 | 1749 | 4223319 | 79 | |
U04299.1 | - | 8 - 9 | 1452 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |