Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008550.2 | NP_032576.2 | 13 - 14 | 2363 | 68 | |
GenBank mRNA | AB006458.1 | - | 13 - 14 | 2341 | 68 | |
AK004900.1 | - | 13 - 14 | 2317 | 68 | ||
AK040388.1 | - | 13 - 14 | 2324 | 68 | ||
AK075649.1 | - | 5 - 6 | 1030 | 68 | ||
AK165376.1 | - | 13 - 14 | 2356 | 68 | ||
BC066211.1 | - | 13 - 14 | 2314 | 30101816 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |