Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK142169.1 | - | 5 - 6 | 892 | 76 | |
AK165778.1 | - | 5 - 6 | 895 | 76 | ||
BC052854.1 | - | 5 - 6 | 894 | 30058638 | 76 | |
Z12604.1 | - | 5 - 6 | 880 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |