Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008638.2 | NP_032664.1 | 5 - 6 | 741 | 84 | |
GenBank mRNA | AK076019.1 | - | 5 - 6 | 694 | 84 | |
AK152249.1 | - | 5 - 6 | 733 | 84 | ||
AK159680.1 | - | 5 - 6 | 714 | 84 | ||
AK165840.1 | - | 5 - 6 | 733 | 84 | ||
AK166262.1 | - | 5 - 6 | 725 | 84 | ||
BC019511.1 | - | 5 - 6 | 728 | 4164253 | 84 | |
J04627.1 | - | 5 - 6 | 684 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |