Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009260.2 | NP_033286.2 | 23 - 24 | 5384 | 101 | |
NM_175836.2 | NP_787030.2 | 23 - 24 | 5044 | 101 | ||
GenBank mRNA | AF017112.1 | - | 23 - 24 | 5291 | 101 | |
AK161119.1 | - | 2 - 3 | 167 | 101 | ||
AK220456.1 | - | 24 - 25 | 5246 | 101 | ||
M74773.1 | - | 23 - 24 | 5364 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |