Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009840.3 | NP_033970.3 | 2 - 3 | 238 | 88 | |
GenBank mRNA | AK010882.1 | - | 2 - 3 | 239 | 88 | |
AK075711.1 | - | 2 - 3 | 219 | 88 | ||
AK075880.1 | - | 2 - 3 | 227 | 88 | ||
AK145661.1 | - | 2 - 3 | 222 | 88 | ||
AK145754.1 | - | 2 - 3 | 221 | 88 | ||
AK145918.1 | - | 2 - 3 | 240 | 88 | ||
AK147038.1 | - | 2 - 3 | 241 | 88 | ||
AK159969.1 | - | 2 - 3 | 230 | 88 | ||
AK168465.1 | - | 2 - 3 | 210 | 88 | ||
AK172867.1 | - | 2 - 3 | 209 | 88 | ||
BC009007.1 | - | 2 - 3 | 196 | 3582639 | 88 | |
Z37164.1 | - | 2 - 3 | 242 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |