Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 16 - 17 | 2143 | 72 | |
GenBank mRNA | AF087436.1 | - | 16 - 17 | 2145 | 72 | |
AK131129.1 | - | 16 - 17 | 2097 | 72 | ||
AK143535.1 | - | 16 - 17 | 2137 | 72 | ||
AK149750.1 | - | 16 - 17 | 2138 | 72 | ||
AK166215.1 | - | 16 - 17 | 2137 | 72 | ||
AK169950.1 | - | 16 - 17 | 2133 | 72 | ||
AK171498.1 | - | 16 - 17 | 2180 | 72 | ||
BC059242.1 | - | 16 - 17 | 2135 | 72 | ||
BC060133.1 | - | 16 - 17 | 2135 | 72 | ||
BC141107.1 | - | 16 - 17 | 2103 | 9055719 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |