Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042557.2 | NP_001036022.1 | 5 - 6 | 663 | 81 | |
NM_001164172.1 | NP_001157644.1 | 5 - 6 | 663 | 81 | ||
NM_001291777.1 | NP_001278706.1 | 4 - 5 | 615 | 81 | ||
NM_001291778.1 | NP_001278707.1 | 4 - 5 | 506 | 81 | ||
NM_001291783.1 | NP_001278712.1 | 4 - 5 | 506 | 81 | ||
NM_011944.3 | NP_036074.2 | 4 - 5 | 615 | 81 | ||
GenBank mRNA | AB005654.1 | - | 5 - 6 | 538 | 81 | |
AF022112.1 | - | 4 - 5 | 592 | 81 | ||
AF022113.1 | - | 5 - 6 | 640 | 81 | ||
AF060943.1 | - | 4 - 5 | 506 | 81 | ||
AF060944.1 | - | 4 - 5 | 526 | 81 | ||
AF060945.1 | - | 4 - 5 | 526 | 81 | ||
AF060946.1 | - | 5 - 6 | 574 | 81 | ||
AF060947.1 | - | 5 - 6 | 574 | 81 | ||
AK165184.1 | - | 4 - 5 | 532 | 81 | ||
AK214564.1 | - | 3 - 4 | 195 | 81 | ||
BC070467.1 | - | 5 - 6 | 575 | 30615302 | 81 | |
U74463.1 | - | 4 - 5 | 472 | 81 | ||
U74464.1 | - | 3 - 4 | 361 | 81 | ||
U93030.1 | - | 4 - 5 | 506 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |