Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013493.3 | NP_038521.1 | 2 - 3 | 339 | 58 | |
GenBank mRNA | AK075760.1 | - | 2 - 3 | 304 | 58 | |
AK144740.1 | - | 2 - 3 | 286 | 58 | ||
AK145444.1 | - | 2 - 3 | 269 | 58 | ||
AK146224.1 | - | 2 - 3 | 280 | 58 | ||
AK146344.1 | - | 2 - 3 | 282 | 58 | ||
AK146367.1 | - | 2 - 3 | 266 | 58 | ||
AK146593.1 | - | 2 - 3 | 255 | 58 | ||
AK151317.1 | - | 2 - 3 | 282 | 58 | ||
AK151528.1 | - | 2 - 3 | 252 | 58 | ||
AK159810.1 | - | 2 - 3 | 256 | 58 | ||
BC058723.1 | - | 2 - 3 | 228 | 5705640 | 58 | |
U20326.1 | - | 2 - 3 | 241 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |