Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290717.1 | NP_001277646.1 | 10 - 11 | 1227 | 75 | |
NM_001290718.1 | NP_001277647.1 | 10 - 11 | 1200 | 75 | ||
NM_001290719.1 | NP_001277648.1 | 10 - 11 | 1227 | 75 | ||
NM_013757.2 | NP_038785.1 | 10 - 11 | 1227 | 75 | ||
GenBank mRNA | AB025258.1 | - | 10 - 11 | 1198 | 75 | |
AB025259.1 | - | 10 - 11 | 1198 | 75 | ||
AB120510.1 | - | 9 - 10 | 1013 | 75 | ||
AK030401.1 | - | 10 - 11 | 1074 | 75 | ||
AK161582.1 | - | 10 - 11 | 1193 | 75 | ||
BC026819.1 | - | 10 - 11 | 1227 | 4488012 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |