Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013909.2 | NP_038937.2 | 5 - 6 | 885 | 72 | |
GenBank mRNA | AB041571.1 | - | 4 - 5 | 1123 | 72 | |
AF176522.1 | - | 5 - 6 | 873 | 72 | ||
AK040458.1 | - | 3 - 4 | 1265 | 72 | ||
BC011485.1 | - | 3 - 4 | 1258 | 72 | ||
BC026818.1 | - | 3 - 4 | 1252 | 72 | ||
BC032263.1 | - | 4 - 5 | 72 | |||
BC044784.2 | - | 4 - 5 | 1085 | 72 | ||
BC054377.1 | - | 4 - 5 | 706 | 6315438 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |