Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016770.3 | NP_058050.3 | 8 - 9 | 1131 | 100 | |
GenBank mRNA | AF026380.1 | - | 8 - 9 | 1113 | 100 | |
AK002920.1 | - | 8 - 9 | 1098 | 100 | ||
AK090289.1 | - | 1132 | 100 | |||
BC119605.1 | - | 8 - 9 | 1075 | 40060386 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |