Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019434.2 | NP_062307.2 | 10 - 11 | 2954 | 93 | |
GenBank mRNA | AJ006590.1 | - | 10 - 11 | 2987 | 93 | |
AK052382.1 | - | 5 - 6 | 810 | 93 | ||
BC052452.1 | - | 10 - 11 | 2954 | 6409827 | 93 | |
BC067414.1 | - | 9 - 10 | 1889 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |