Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019481.2 | NP_062354.2 | 5 - 6 | 641 | 134 | |
GenBank mRNA | AF199365.1 | - | 5 - 6 | 641 | 134 | |
AF199366.1 | - | 5 - 6 | 641 | 134 | ||
AK128899.1 | - | 5 - 6 | 641 | 134 | ||
BC040789.1 | - | 5 - 6 | 615 | 134 | ||
BC049981.1 | - | 5 - 6 | 623 | 5065026 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |