Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019579.3 | NP_062525.1 | 13 - 14 | 2147 | 81 | |
GenBank mRNA | AF199008.1 | - | 13 - 14 | 2078 | 81 | |
AK133187.1 | - | 13 - 14 | 2040 | 81 | ||
BC067404.1 | - | 13 - 14 | 2008 | 81 | ||
BC075698.1 | - | 13 - 14 | 2100 | 81 | ||
BC138624.1 | - | 13 - 14 | 2026 | 8861644 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |