Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019749.4 | NP_062723.1 | 1 - 2 | 433 | 79 | |
GenBank mRNA | AF161587.1 | - | 1 - 2 | 153 | 79 | |
AK002879.1 | - | 1 - 2 | 188 | 79 | ||
AK011731.1 | - | 1 - 2 | 98 | 79 | ||
AV029091.2 | - | 1 - 2 | 204 | 79 | ||
BC002126.1 | - | 1 - 2 | 196 | 3497484 | 79 | |
BC024621.1 | - | 1 - 2 | 216 | 4459651 | 79 | |
BC029329.1 | - | 1 - 2 | 135 | 79 | ||
BC030350.1 | - | 1 - 2 | 442 | 5102114 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |