Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rec8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020002.3 | NP_064386.2 | 18 - 19 | 1824 | 61 | |
GenBank mRNA | AF262055.1 | - | 18 - 19 | 1754 | 61 | |
AK077167.1 | - | 18 - 19 | 1759 | 61 | ||
AK146832.1 | - | 18 - 19 | 1810 | 61 | ||
BC052155.1 | - | 18 - 19 | 1820 | 6335959 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |