Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020568.3 | NP_065593.2 | 6 - 7 | 3738 | 65 | |
GenBank mRNA | AF064748.1 | - | 6 - 7 | 3749 | 65 | |
AK173286.1 | - | 6 - 7 | 2470 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |