Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020594.2 | NP_065619.2 | 3 - 4 | 505 | 106 | |
GenBank mRNA | AF061961.1 | - | 3 - 4 | 513 | 106 | |
AK132679.1 | - | 3 - 4 | 452 | 106 | ||
AK164491.1 | - | 3 - 4 | 452 | 106 | ||
BC048687.1 | - | 3 - 4 | 437 | 6334861 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |