Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021339.2 | NP_067314.2 | 1 - 2 | 109 | 97 | |
GenBank mRNA | AF090866.1 | - | 1 - 2 | 109 | 97 | |
AK040711.1 | - | 2 - 3 | 201 | 97 | ||
AK157750.1 | - | 3 - 4 | 292 | 97 | ||
BC109176.2 | - | 1 - 2 | 108 | 40048829 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |