Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021790.2 | NP_068562.1 | 7 - 8 | 772 | 110 | |
NM_181061.5 | NP_851406.1 | 7 - 8 | 895 | 110 | ||
NR_075088.1 | 7 - 8 | 908 | 110 | |||
GenBank mRNA | AB043687.1 | - | 7 - 8 | 772 | 110 | |
AK082939.1 | - | 7 - 8 | 552 | 110 | ||
AK135773.1 | - | 288 | 110 | |||
AK160776.1 | - | 7 - 8 | 543 | 110 | ||
BC028655.1 | - | 7 - 8 | 908 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |