Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022012.3 | NP_071295.2 | 9 - 10 | 2472 | 101 | |
GenBank mRNA | AK159518.1 | - | 7 - 8 | 1492 | 101 | |
BC030928.1 | - | 7 - 8 | 1025 | 101 | ||
BC047152.1 | - | 7 - 8 | 2219 | 6476975 | 101 | |
BC095963.1 | - | 9 - 10 | 2472 | 5400993 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |