Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2159309 |
BC158045.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2159309 |
NM_080451.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095133 |
AK075917.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095133 |
NM_080451.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
AK079194.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
BC158045.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
NM_080451.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
AJ306625.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
AK079194.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
BC158045.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
NM_080451.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P223127 |
AJ306625.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
943207 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
733498 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
721028 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
382458 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
46016 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334086 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626198 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
80950 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532144 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
955054 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546665 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674233 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374267 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
440593 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600735 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
874630 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154877 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
192725 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347560 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
219429 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153514 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57787 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301728 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
566248 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669881 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703261 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
127729 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518136 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
127574 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
271518 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
466747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640141 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
215424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344466 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
95846 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545303 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
38285 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54474 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
263766 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
660063 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
664280 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633434 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453600 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
813923 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
81680 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
617928 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
105562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
958457 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
121809 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
82911 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
247658 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702421 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931906 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
152984 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
22130 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240692 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842597 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
486226 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
424979 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
387714 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798068 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
625583 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354723 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
507649 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
745224 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
941539 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
765644 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
624143 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598408 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
425913 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808217 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579794 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
343852 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469657 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
938977 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9477 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
19886 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
768062 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
296342 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554534 |
AK035191.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554534 |
AK045406.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554534 |
BC027158.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554534 |
BC049153.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
606559 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325432 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AF119383.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AF352245.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AK034121.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AK050384.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AK079180.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AK141839.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AK165081.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
AY290823.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
BC106840.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
BC106841.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386868 |
NM_033622.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
499116 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
705696 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
420112 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
477739 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45453 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770223 |
BC119031.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770223 |
D49921.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770223 |
D88264.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770223 |
NM_010275.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770223 |
U37459.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770223 |
U75532.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
130674 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658009 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305454 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
33144 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633577 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
221210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187887 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
934436 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616323 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371635 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
32231 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162553 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
22693 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
150189 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
79295 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157586 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
1305 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178955 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
123681 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
97940 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
169531 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
24297 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
119070 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
86082 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
163945 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
180515 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
23815 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
21484 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
40255 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
AK079194.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
BC158045.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
NM_080451.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
AJ306625.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
AK079194.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
BC158045.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
NM_080451.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P223127 |
AJ306625.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2159309 |
BC158045.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2159309 |
NM_080451.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095133 |
AK075917.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095133 |
NM_080451.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P948080 |
AK075917.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P948080 |
NM_080451.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P119776 |
AK004418.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
AK079194.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
BC158045.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
NM_080451.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
AJ306625.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
AK079194.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
BC158045.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
NM_080451.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P223127 |
AJ306625.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P138863 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486027 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486053 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486011 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486058 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486311 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486259 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486264 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02726807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486036 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02553887 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486038 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486239 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486063 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486093 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486210 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02719792 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486261 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02793946 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486285 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02793950 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486274 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486266 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486278 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02326236 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486295 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486242 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486010 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02921037 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486067 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486317 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486253 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486301 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486033 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486003 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486289 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486057 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486268 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486245 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486004 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02864639 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486241 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486214 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02726804 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03189209 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486059 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02719802 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486312 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486275 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02726802 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486280 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486309 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486049 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486082 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486008 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486212 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486299 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486288 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486025 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486062 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486030 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486086 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486255 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02719801 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03091917 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03042418 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486291 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486247 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02793944 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02726803 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486209 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02726808 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486029 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486006 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486217 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03200373 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486216 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486055 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486079 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486065 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486077 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03091926 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486243 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486277 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486251 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486070 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486279 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486307 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486054 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486258 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02328501 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486314 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02518128 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486061 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486270 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22486009 |
- |
- |