Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008166.2 | NP_032192.2 | 12 - 13 | 2091 | 83 | |
GenBank mRNA | AK138279.1 | - | 12 - 13 | 2081 | 83 | |
D10171.1 | - | 12 - 13 | 2061 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |