Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281980.1 | NP_001268909.1 | 4 - 5 | 376 | 92 | |
NM_001281981.1 | NP_001268910.1 | 5 - 6 | 417 | 92 | ||
NM_008511.3 | NP_032537.3 | 4 - 5 | 648 | 92 | ||
GenBank mRNA | AK040180.1 | - | 4 - 5 | 649 | 92 | |
AK079932.1 | - | 4 - 5 | 606 | 92 | ||
AK153823.1 | - | 4 - 5 | 646 | 92 | ||
AK153865.1 | - | 4 - 5 | 378 | 92 | ||
AK160009.1 | - | 4 - 5 | 608 | 92 | ||
AK169578.1 | - | 5 - 6 | 401 | 92 | ||
AY753324.1 | - | 3 - 4 | 211 | 92 | ||
BC052909.1 | - | 4 - 5 | 584 | 30076553 | 92 | |
U10484.1 | - | 4 - 5 | 354 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |