Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009054.3 | NP_033080.2 | 4 - 5 | 1338 | 63 | |
GenBank mRNA | AK031744.1 | - | 4 - 5 | 1056 | 63 | |
AK088180.1 | - | 4 - 5 | 822 | 63 | ||
BC003219.1 | - | 4 - 5 | 843 | 3482042 | 63 | |
BC069924.1 | - | 4 - 5 | 1049 | 63 | ||
BC085503.1 | - | 4 - 5 | 1045 | 63 | ||
CA549428.1 | - | 4 - 5 | 389 | 63 | ||
L46855.1 | - | 949 | 63 | |||
X75343.1 | - | 4 - 5 | 753 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |