Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009372.3 | NP_033398.2 | 1 - 2 | 344 | 76 | |
GenBank mRNA | AK008909.1 | - | 1 - 2 | 346 | 76 | |
BC005724.1 | - | 1 - 2 | 343 | 3595273 | 76 | |
CD563532.1 | - | 1 - 2 | 260 | 76 | ||
X89749.1 | - | 1 - 2 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons |