Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009475.2 | NP_033501.2 | 1 - 2 | 72 | 72 | |
GenBank mRNA | BC051092.1 | - | 1 - 2 | 60 | 6529097 | 72 |
U28486.1 | - | 1 - 2 | 71 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |