Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009509.2 | NP_033535.2 | 13 - 14 | 1560 | 80 | |
GenBank mRNA | AK027908.1 | - | 13 - 14 | 1557 | 80 | |
BC015267.1 | - | 13 - 14 | 1536 | 4236751 | 80 | |
BC117875.2 | - | 13 - 14 | 1525 | 40095718 | 80 | |
BC145671.1 | - | 13 - 14 | 1527 | 40130974 | 80 | |
M98454.1 | - | 13 - 14 | 1550 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |