Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289765.1 | NP_001276694.1 | 3 - 4 | 763 | 79 | |
NM_001289766.1 | NP_001276695.1 | 3 - 4 | 728 | 79 | ||
NM_009539.3 | NP_033565.2 | 3 - 4 | 720 | 79 | ||
GenBank mRNA | AK039883.1 | - | 3 - 4 | 657 | 79 | |
AK089056.1 | - | 3 - 4 | 693 | 79 | ||
BC029727.1 | - | 3 - 4 | 678 | 4925739 | 79 | |
U04379.1 | - | 3 - 4 | 674 | 79 | ||
U77667.1 | - | 2 - 3 | 566 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |