Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010280.3 | NP_034410.3 | 3 - 4 | 632 | 95 | |
GenBank mRNA | AB008833.1 | - | 3 - 4 | 510 | 95 | |
AF020305.1 | - | 3 - 4 | 465 | 95 | ||
AF036163.1 | - | 3 - 4 | 526 | 95 | ||
AF041842.1 | - | 3 - 4 | 515 | 95 | ||
AF051766.1 | - | 3 - 4 | 527 | 95 | ||
AK046542.1 | - | 3 - 4 | 559 | 95 | ||
BC066202.1 | - | 3 - 4 | 632 | 6430904 | 95 | |
Y15110.1 | - | 3 - 4 | 546 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |