Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010357.3 | NP_034487.2 | 3 - 4 | 257 | 109 | |
GenBank mRNA | AK008189.1 | - | 3 - 4 | 243 | 109 | |
AK008193.1 | - | 4 - 5 | 208 | 109 | ||
AK008400.1 | - | 3 - 4 | 241 | 109 | ||
AK008490.1 | - | 3 - 4 | 208 | 109 | ||
AK009668.1 | - | 3 - 4 | 211 | 109 | ||
AK010098.1 | - | 3 - 4 | 211 | 109 | ||
AK011177.1 | - | 3 - 4 | 208 | 109 | ||
AK011841.1 | - | 3 - 4 | 236 | 109 | ||
AK019271.1 | - | 3 - 4 | 232 | 109 | ||
BC012639.1 | - | 3 - 4 | 211 | 3995378 | 109 | |
BU953110.1 | - | 3 - 4 | 260 | 109 | ||
L06047.1 | - | 3 - 4 | 209 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |