Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136067.2 | NP_001129539.1 | 6 - 7 | 937 | 55 | |
NM_001161421.1 | NP_001154893.1 | 6 - 7 | 934 | 55 | ||
NM_001161422.1 | NP_001154894.1 | 5 - 6 | 755 | 55 | ||
NM_001161423.1 | NP_001154895.1 | 5 - 6 | 807 | 55 | ||
NM_001161424.1 | NP_001154896.1 | 5 - 6 | 804 | 55 | ||
NM_010547.2 | NP_034677.2 | 5 - 6 | 833 | 55 | ||
NM_178590.4 | NP_848705.1 | 5 - 6 | 830 | 55 | ||
GenBank mRNA | AF069542.1 | - | 4 - 5 | 692 | 55 | |
AF513109.1 | - | 6 - 7 | 848 | 55 | ||
AK037020.1 | - | 5 - 6 | 809 | 55 | ||
AK148163.1 | - | 5 - 6 | 791 | 55 | ||
AK154095.1 | - | 5 - 6 | 819 | 55 | ||
AY112937.1 | - | 6 - 7 | 937 | 55 | ||
BC021431.1 | - | 6 - 7 | 924 | 5030381 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |