Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159577.1 | NP_001153049.1 | 3 - 4 | 831 | 55 | |
NM_001159578.1 | NP_001153050.1 | 4 - 5 | 976 | 55 | ||
NM_001159579.1 | NP_001153051.1 | 2 - 3 | 607 | 55 | ||
NM_001159580.1 | NP_001153052.1 | 2 - 3 | 607 | 55 | ||
NM_010727.4 | NP_034857.3 | 2 - 3 | 607 | 55 | ||
GenBank mRNA | AF034745.1 | - | 3 - 4 | 831 | 55 | |
AF034746.1 | - | 2 - 3 | 535 | 55 | ||
AK044127.1 | - | 2 - 3 | 560 | 55 | ||
AK138326.1 | - | 2 - 3 | 608 | 55 | ||
AK161194.1 | - | 4 - 5 | 962 | 55 | ||
BC040367.1 | - | 2 - 3 | 559 | 55 | ||
BC139771.1 | - | 2 - 3 | 535 | 40111506 | 55 | |
KJ418422.1 | - | 2 - 3 | 305 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |