Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011539.3 | NP_035669.3 | 1 - 2 | 253 | 73 | |
GenBank mRNA | AK048544.1 | - | 1 - 2 | 235 | 73 | |
AK139479.1 | - | 1 - 2 | 216 | 73 | ||
AK162394.1 | - | 358 | 73 | |||
BC029014.1 | - | 1 - 2 | 255 | 5007629 | 73 | |
L18868.1 | - | 1 - 2 | 124 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |