Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290712.1 | NP_001277641.1 | 5 - 6 | 821 | 85 | |
NM_013686.4 | NP_038714.2 | 6 - 7 | 907 | 85 | ||
GenBank mRNA | AK149611.1 | - | 6 - 7 | 772 | 85 | |
AK149755.1 | - | 6 - 7 | 773 | 85 | ||
AK150666.1 | - | 4 - 5 | 2332 | 85 | ||
AK150744.1 | - | 4 - 5 | 2332 | 85 | ||
AK151068.1 | - | 6 - 7 | 772 | 85 | ||
AK151445.1 | - | 5 - 6 | 711 | 85 | ||
AK151903.1 | - | 4 - 5 | 2332 | 85 | ||
AK152633.1 | - | 4 - 5 | 2332 | 85 | ||
AK152641.1 | - | 6 - 7 | 764 | 85 | ||
AK152759.1 | - | 6 - 7 | 773 | 85 | ||
AK153037.1 | - | 6 - 7 | 773 | 85 | ||
AK153430.1 | - | 5 - 6 | 711 | 85 | ||
AK165665.1 | - | 6 - 7 | 774 | 85 | ||
AK166828.1 | - | 6 - 7 | 771 | 85 | ||
AK166966.1 | - | 6 - 7 | 795 | 85 | ||
BC003809.1 | - | 6 - 7 | 755 | 3599670 | 85 | |
D90344.1 | - | 6 - 7 | 722 | 85 | ||
M12899.1 | - | 6 - 7 | 730 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |