Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013806.2 | NP_038834.2 | 12 - 13 | 1758 | 83 | |
GenBank mRNA | AF227274.1 | - | 12 - 13 | 1662 | 83 | |
AF282772.1 | - | 12 - 13 | 1662 | 83 | ||
AF282773.1 | - | 12 - 13 | 1662 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |