Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il27ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016671.3 | NP_057880.2 | 12 - 13 | 1643 | 140 | |
GenBank mRNA | AF053005.1 | - | 12 - 13 | 1611 | 140 | |
AF265243.1 | - | 12 - 13 | 1622 | 140 | ||
AK040457.1 | - | 10 - 11 | 1983 | 140 | ||
BC032878.1 | - | 12 - 13 | 1643 | 1244998 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |