Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038710.1 | NP_001033799.1 | 4 - 5 | 839 | 77 | |
NM_016711.3 | NP_057920.2 | 4 - 5 | 775 | 77 | ||
GenBank mRNA | AF177170.1 | - | 4 - 5 | 724 | 77 | |
AF237629.1 | - | 5 - 6 | 683 | 77 | ||
AK015263.1 | - | 4 - 5 | 820 | 77 | ||
AK015719.1 | - | 4 - 5 | 875 | 77 | ||
AK018223.1 | - | 4 - 5 | 572 | 77 | ||
AK076562.1 | - | 4 - 5 | 789 | 77 | ||
AK079077.1 | - | 4 - 5 | 619 | 77 | ||
AK147581.1 | - | 4 - 5 | 771 | 77 | ||
AK164178.1 | - | 4 - 5 | 621 | 77 | ||
BC052227.1 | - | 4 - 5 | 839 | 77 | ||
BC061124.1 | - | 4 - 5 | 605 | 6531518 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |