Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016712.3 | NP_057921.1 | 3 - 4 | 386 | 70 | |
GenBank mRNA | AF177174.1 | - | 2 - 3 | 305 | 70 | |
AK144408.1 | - | 3 - 4 | 387 | 70 | ||
AK144535.1 | - | 3 - 4 | 387 | 70 | ||
BC068020.1 | - | 2 - 3 | 322 | 30291625 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |