Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016963.2 | NP_058659.1 | 1 - 2 | 160 | 89 | |
GenBank mRNA | AF177172.1 | - | 1 - 2 | 148 | 89 | |
AF237630.1 | - | 1 - 2 | 74 | 89 | ||
AK017725.1 | - | 1 - 2 | 157 | 89 | ||
AK050372.1 | - | 1 - 2 | 162 | 89 | ||
AK082971.1 | - | 1 - 2 | 161 | 89 | ||
AK147017.1 | - | 1 - 2 | 161 | 89 | ||
AK167723.1 | - | 1 - 2 | 158 | 89 | ||
AK169218.1 | - | 1 - 2 | 161 | 89 | ||
BC082595.1 | - | 1 - 2 | 152 | 30645500 | 89 | |
BY067594.1 | - | 1 - 2 | 161 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |