Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019663.3 | NP_062637.2 | 3 - 4 | 559 | 71 | |
GenBank mRNA | AF077950.1 | - | 3 - 4 | 551 | 71 | |
AK075708.1 | - | 3 - 4 | 560 | 71 | ||
AK153871.1 | - | 3 - 4 | 564 | 71 | ||
BC051417.1 | - | 3 - 4 | 556 | 6515027 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |