Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199118.1 | NP_001186047.1 | 8 - 9 | 1448 | 69 | |
NM_022883.3 | NP_075021.1 | 8 - 9 | 1525 | 69 | ||
GenBank mRNA | AF286724.1 | - | 8 - 9 | 1462 | 69 | |
AK033647.1 | - | 8 - 9 | 1446 | 69 | ||
AK049363.1 | - | 8 - 9 | 1441 | 69 | ||
AK163833.1 | - | 8 - 9 | 1440 | 69 | ||
AK220228.1 | - | 8 - 9 | 1525 | 69 | ||
BC026729.1 | - | 8 - 9 | 1432 | 69 | ||
BC117882.1 | - | 7 - 8 | 1389 | 40096038 | 69 | |
BC117883.1 | - | 7 - 8 | 1389 | 40096041 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |