Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 8 - 9 | 806 | 65 | |
NM_026653.2 | NP_080929.1 | 7 - 8 | 743 | 65 | ||
GenBank mRNA | AK019864.1 | - | 1 - 2 | 144 | 65 | |
AK035084.1 | - | 7 - 8 | 671 | 65 | ||
AK148035.1 | - | 7 - 8 | 684 | 65 | ||
AK150785.1 | - | 7 - 8 | 640 | 65 | ||
AK151590.1 | - | 7 - 8 | 696 | 65 | ||
AK152296.1 | - | 8 - 9 | 736 | 65 | ||
AK153107.1 | - | 8 - 9 | 736 | 65 | ||
AK153144.1 | - | 7 - 8 | 696 | 65 | ||
AK165316.1 | - | 7 - 8 | 656 | 65 | ||
AK165944.1 | - | 7 - 8 | 674 | 65 | ||
AK167598.1 | - | 7 - 8 | 629 | 65 | ||
AK169599.1 | - | 7 - 8 | 667 | 65 | ||
AK216917.1 | - | 2 - 3 | 202 | 65 | ||
BC019119.1 | - | 7 - 8 | 674 | 4487586 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |