Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025951.3 | NP_080227.2 | 3 - 4 | 736 | 99 | |
NM_028744.3 | NP_083020.2 | 3 - 4 | 667 | 99 | ||
GenBank mRNA | AK016754.1 | - | 3 - 4 | 633 | 99 | |
AK035523.1 | - | 3 - 4 | 721 | 99 | ||
AK170730.1 | - | 3 - 4 | 720 | 99 | ||
AY148879.1 | - | 633 | 99 | |||
BC062144.1 | - | 3 - 4 | 643 | 6335661 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |