Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053273.2 | NP_444503.2 | 8 - 9 | 1000 | 65 | |
GenBank mRNA | AF329682.1 | - | 8 - 9 | 943 | 65 | |
AK152172.1 | - | 8 - 9 | 952 | 65 | ||
AK154835.1 | - | 8 - 9 | 1008 | 65 | ||
AK155166.1 | - | 8 - 9 | 1001 | 65 | ||
AK156393.1 | - | 8 - 9 | 953 | 65 | ||
AK168408.1 | - | 8 - 9 | 953 | 65 | ||
AK204126.1 | - | 1 - 2 | 42 | 65 | ||
BC060740.1 | - | 8 - 9 | 943 | 4158813 | 65 | |
BY731753.1 | - | 4 - 5 | 311 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |