Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130452.1 | NP_569719.1 | 2 - 3 | 431 | 85 | |
GenBank mRNA | AK050010.1 | - | 449 | 85 | ||
AY033514.1 | - | 2 - 3 | 369 | 85 | ||
BC019406.1 | - | 2 - 3 | 431 | 5134018 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |