Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cux2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007804.2 | NP_031830.2 | 7 - 8 | 828 | 60 | |
GenBank mRNA | BC065113.1 | - | 7 - 8 | 828 | 30532644 | 60 |
U45665.1 | - | 7 - 8 | 649 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |