Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008777.3 | NP_032803.2 | 11 - 12 | 1417 | 86 | |
GenBank mRNA | AK145839.1 | - | 11 - 12 | 1282 | 86 | |
AK149519.1 | - | 11 - 12 | 1280 | 86 | ||
BC013458.1 | - | 11 - 12 | 1275 | 4223155 | 86 | |
X51942.1 | - | 11 - 12 | 1242 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |